EMAIL THIS PAGE TO A FRIEND
Z741071 Sigma
Go Matrix, 6 mm
pore size 130 μm, vial of 48 ea
Purchase
Properties
Related Categories 3D Cell Culture Products, 3D Scaffolds, Cell Culture Supplies, Gelatin Scaffold: Go Matrix gelatin based scaffolds, Labware More...
packaging   vial of 48 ea
thickness   1.5 mm
pore size   130 μm
Description
Features and Benefits
• Made of easily manipulated material
• Custom pore size and porosity
• Uniform micro-environment
• Open for cell communication
General description
3D Cell Culture Gelatin Scaffold
Go Matrix is a uniform micro-environment for 3D culture modeling. Cells form 3D morphology in uniform pores of Go Matrix.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?