• USA Home
  • Z741613 - Prospenser Bottle-top Dispenser
EMAIL THIS PAGE TO A FRIEND
Z741613
Prospenser Bottle-top Dispenser
dispensing volume 0.01-2.5 mL
  •  NACRES NB.01
Purchase
Properties
Related Categories Bottletop dispensers, Dispensers, Labware, Liquid Handling Equipment, Manufacturer Browser,
feature   with recirculating valve
mfr. no.   Sartorius, 723049
dispensing volume measuring range   0.01-2.5 mL
volume   , Adjustable in 0.05mL increments
  0.01-2.5 mL
fitting   (38mm)
  (40 mm)
  45 mm
Description
General description
The Prospenser bottle-top dispensers deliver safe and precise dispensing of liquids, including strong acids, bases and solvents from a supply bottle. Special features include an easy-to-adjust calibration mechanism and anti-drip precision valve for enhanced accuracy and usability. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Prospensers are fully autoclavable and easy to dismantle for cleaning and maintenance. Wide range of adapters are included to fit most common bottle sizes. All models are fitted with an anti-drip safety valve or a closure cap, a feature which guards against drips, when the bottle-top dispensers are not in use. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Wide range of adapters included to fit most common bottle sizes. Fully autoclavable at 121o C, 2 bar for 20 minutes.The Prospenser bottle-top dispensers deliver safe and precise dispensing of liquids, including strong acids, bases and solvents from a supply bottle. Special features include an easy-to-adjust calibration mechanism and anti-drip precision valve for enhanced accuracy and usability. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Prospensers are fully autoclavable and easy to dismantle for cleaning and maintenance. Wide range of adapters are included to fit most common bottle sizes. All models are fitted with an anti-drip safety valve or a closure cap, a feature which guards against drips, when the bottle-top dispensers are not in use. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Wide range of adapters included to fit most common bottle sizes. Fully autoclavable at 121o C, 2 bar for 20 minutes.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?