• USA Home
  • Z759368 - Nanofiber multiwell plate, Interactive MicroEnvironMent System (IMEMS)

EMAIL THIS PAGE TO A FRIEND
Z759368 Sigma-Aldrich

Nanofiber multiwell plate, Interactive MicroEnvironMent System (IMEMS)

size 6 wells, for co-culture, aligned nanofibers

Synonym: 3D plates, tissue culture plates

Purchase

Properties

Related Categories 3D Cell Culture Products, 3D Scaffolds, Cell Culture Supplies, Labware, Nanofiber Scaffolds More...
material   clear wells flat
  transparent polycaprolactone (PCL) plate
sterility   sterile
feature   lid
mfr. no.   Nanofiber Solutions, IMEM02
size   6 wells
color   clear
  transparent
matrix active group   Non-treated surface treatment
suitability   suitable for (Tissue Engineering, Cancer Research)
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?