• USA Home
  • 06271501 - Vanilla extract, synthetic

EMAIL THIS PAGE TO A FRIEND
06271501

Vanilla extract, synthetic

secondary reference standard

Purchase

Description

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Vanillin, Vanillic acid (quantitative); Vanillin, Vanillic acid, Ethylvanillin (qualitative)

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Vanilla – Natural or Out of the Reaction Flask

Vanilla is one of the most popular flavors in food and beverage products. The demand far exceeds the global supply of naturally grown vanilla; therefore, in addition to natural vanilla, artificial va...
Keywords: Food & Beverage, High performance liquid chromatography, Indicators, Sample preparations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?