EMAIL THIS PAGE TO A FRIEND
11872 Supelco

Coated GC Packing

phase 10% SP-1000, matrix 80/100 SUPELCOPORT support, bottle of 20 g

Purchase

Properties

packaging   bottle of 20 g
parameter   50-250 °C temp. range
matrix   80/100 SUPELCOPORT support
matrix active group   10% SP-1000 phase
compatibility   for analyte group general anaerobic fermentation

Description

General description

Ready-to-use coated packing for those wanting to pack their own columns.

Application

general use

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?