• USA Home
  • 12543-U - Metal Packed GC Column (General Configuration)

EMAIL THIS PAGE TO A FRIEND
12543-U Supelco

Metal Packed GC Column (General Configuration)

phase 1% SP-1000, matrix 60/80 Carbopack B support, L × O.D. × I.D. 8.0 ft (2.4 m) × 1/8  in. × 2.1 mm

  •  eCl@ss 32110502

  •  NACRES SB.54

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (General Configuration), Packed GC Columns, Packed GC Columns and Components More...
Quality Level   100
material   stainless steel column
parameter   0-225 °C temperature (isothermal or programmed)
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   8.0 ft (2.4 m) × 1/8  in. × 2.1 mm
matrix   60/80 Carbopack B support
matrix active group   1% SP-1000 phase
column type   packed GC

Description

General description

Packed column made up of stainless steel are basically employed when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents.

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.

Application

Carbopack B SP-1000 packed column maybe used for the quantification of synthetic organic chemicals in biological treatment effluent process.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?