• USA Home
  • 12712 - Metal Packed GC Column (General Configuration)

EMAIL THIS PAGE TO A FRIEND
12712 Supelco

Metal Packed GC Column (General Configuration)

phase none, matrix 80/100 Chromosorb 101 support, L × O.D. × I.D. 6.0 ft (1.83 m) × 1/8  in. × 2.1 mm

  •  eCl@ss 32110502

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (General Configuration), Packed GC Columns, Packed GC Columns and Components More...
material   stainless steel column
parameter   0-250 °C temperature (isothermal or programmed)
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   6.0 ft (1.83 m) × 1/8  in. × 2.1 mm
matrix   80/100 Chromosorb 101 support
matrix active group   none phase
column type   packed GC

Description

General description

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?