• USA Home
  • 13027-U - Carbon Adsorbent Sampler Kit

EMAIL THIS PAGE TO A FRIEND
13027-U Supelco

Carbon Adsorbent Sampler Kit

matrix 20/40 GCB Kit

  •  eCl@ss 32161000

  •  NACRES SB.54

Purchase

Properties

Related Categories Adsorbents, Analytical/Chromatography, Carbon Adsorbent Sampler Kits, Gas Chromatography (GC), Scalable Specialty Carbon Adsorbents,
Quality Level   100
packaging   kit of 5 bottles
matrix   20/40 GCB Kit
  Graphitized Carbon Black (GCB) adsorbents
particle size   20-40 mesh

Description

General description

Choosing the right adsorbent or combination of adsorbents can be difficult:
• Must choose one or more to retain specific analyte(s)
• May be equally important to release analyte(s) during desorption

This convenient kit allows a cost-effective way to evaluate several of our specialty carbon adsorbents. Once you find an adsorbent or adsorbents that serve your purpose, we will work with you to either make your device for you, or to supply you bulk adsorbent (in a bottle, jar, or ampule; not packed inside a column or device).

For more information about any of our specialty carbon adsorbents, please visit sigma-aldrich.com/carbon

Other Notes

Purchase of this kit is limited to two per customer per calendar year.

Footnote

Each bottle contains 5 g of a different material.

Kit component only

Description

Product #

Add to Cart

20/40 Carbotrap® B 5 g    
20/40 Carbotrap C 5 g    
20/40 Carbotrap F 5 g    
20/40 Carbotrap X 5 g    
20/40 Carbotrap Y 5 g    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Adsorbent Selection Guide | Technical Article

General Guidelines for Choosing Carbon Adsorbents Physical Characteristics of Supelco Carbon Adsorbents Adsorbent Cross Reference
Keywords: Gas chromatography

Carbon Adsorbents: Physical Characteristics and Selection Guide

The following information is gathered to assist chemists and method developers in locating the proper specialty carbon adsorbent for their use. Additional assistance can be obtained by contacting our...
Keywords: Adsorption, Air Monitoring, Gas chromatography

High Purity Carbon Adsorbents for Sample Preparation and Chromatographic Applications

The first high purity Supelco® carbon adsorbents were made for gas chromatographic packed columns. Then, specialty carbons were developed for thermal desorption tubes followed by the introduction of ...
Keywords: Adsorption, Air sampling, Chromatography, Diffusion, Flame ionization detector, Gas chromatography, High performance liquid chromatography, Passive sampling, Pesticides, Phase transitions, Purification, Sample preparations, Separation, Solid phase extractions, Solid phase microextractions, Thermal conductivity detector

Scalable Specialty Carbon Adsorbents for Purification and Recovery Applications

What is a “Specialty Carbon Adsorbent”? A graphic depicting a spherical particle is shown in Figure 1. Controlling pore composition is very important as it determines the adsorption and desorption ch...
William R. Betz, Michael J. Keeler, Michael D. Buchanan
Reporter US Volume 33.2
Keywords: Adsorption, Environmental, Industries, Oxidations, Petrochemical, Pharmaceutical, Purification, Solvents

Related Content

Choosing the Right Adsorbent for Your Thermal Desorption Gas Chromatography Applications [VIDEO]

This webinar provides an overview of adsorbents that are suitable for thermal desorption (TD), and the factors you need to consider when selecting an adsorbent. The presentation provides details abou...
Keywords: Sample preparations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?