• USA Home
  • 13037-U - Metal Packed GC Column (General Configuration)

EMAIL THIS PAGE TO A FRIEND
13037-U Supelco

Metal Packed GC Column (General Configuration)

phase none, matrix 80/100 Porapak Q support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8  in. × 2.1 mm

  •  NACRES SB.54

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (General Configuration), Packed GC Columns, Packed GC Columns and Components More...
Quality Level   100
material   stainless steel column
parameter   0-250 °C temperature (isothermal or programmed)
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   6.0 ft (1.8 m) × 1/8  in. × 2.1 mm
matrix   80/100 Porapak Q support
matrix active group   none phase
column type   packed GC

Description

General description

The carrier gas percolates through the porous bed in packed columns. In a packed column, separation is achieved by moving the solutes apart. Packed columns are made up of mainly two materials; stainless steel and Pyrex glass. Metal packed columns are more suitable for long and high carrier gas flow rates to tolerate the elevated pressures. Porapak Q is a porous copolymer of styrene and ethylvinylbenzene crosslinked with divinylbenzene which is applicable for quantitative adsorption.

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
What is the maximum temperature for Product No. 13037-U Porapak-Q packed column?
The maximum temperature is 250 deg C.
What is the difference between Stainless Steel Packed Columns Product No. 13037-U and Product No. 12437
The columns are identical except for the fittings shipped with them.Product No. 13037-U is shipped with stainless steel nuts and M-2A ferrules.Product No. 12437 is shipped with brass nuts and ferrules.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?