• USA Home
  • 13384-U - Carbon Adsorbent Sampler Kit
EMAIL THIS PAGE TO A FRIEND
13384-U Supelco
Carbon Adsorbent Sampler Kit
matrix SGPC Kit
  •  NACRES SB.54
Purchase
Properties
Related Categories Adsorbents, Analytical/Chromatography, Carbon Adsorbent Sampler Kits, Gas Chromatography (GC), Specialty Carbon Adsorbents More...
Quality Level   100
packaging   kit of 4 bottles
matrix   SGPC Kit
  Spherical Graphitized Polymer Carbon (SGPC) adsorbents
particle size   (various mesh)
Description
General description
Choosing the right adsorbent or combination of adsorbents can be difficult:
• Must choose one or more to retain specific analyte(s)
• May be equally important to release analyte(s) during desorption
This convenient kit allows a cost-effective way to evaluate several of our specialty carbon adsorbents. Once you find an adsorbent or adsorbents that serve your purpose, we will work with you to either make your device for you, or to supply you bulk adsorbent (in a bottle, jar, or ampule; not packed inside a column or device).
For more information about any of our specialty carbon adsorbents, please visit sigma-aldrich.com/carbon
Other Notes
Purchase of this kit is limited to two per customer per calendar year.
Footnote
Each bottle contains 5 g of a different material.
Kit component only
Description
Product #
Add to Cart
60/80 Graphsphere 2016 5 g    
30/45 Graphsphere 2017 5 g    
50-60 μm Graphsphere 2027 5 g    
120/400 Graphsphere 2029 5 g    
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
Carbon Adsorbents: Physical Characteristics and Selection Guide
The following information is gathered to assist chemists and method developers in locating the proper specialty carbon adsorbent for their use. Additional assistance can be obtained by contacting our...
Keywords: Adsorption, Air Monitoring, Gas chromatography
High Purity Carbon Adsorbents for Sample Preparation and Chromatographic Applications
The first high purity Supelco® carbon adsorbents were made for gas chromatographic packed columns. Then, specialty carbons were developed for thermal desorption tubes followed by the introduction of ...
Keywords: Adsorption, Air sampling, Chromatography, Diffusion, Flame ionization detector, Gas chromatography, High performance liquid chromatography, Passive sampling, Pesticides, Phase transitions, Purification, Sample preparations, Separation, Solid phase extractions, Solid phase microextractions, Thermal conductivity detector
Related Content
Choosing the Right Adsorbent for Your Thermal Desorption Gas Chromatography Applications [VIDEO]
This webinar provides an overview of adsorbents that are suitable for thermal desorption (TD), and the factors you need to consider when selecting an adsorbent. The presentation provides details abou...
Keywords: Sample preparations
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?