• USA Home
  • 13984-U - Metal Packed GC Column (General Configuration)

EMAIL THIS PAGE TO A FRIEND
13984-U Supelco

Metal Packed GC Column (General Configuration)

phase 30% DC-200 (500 cstks), matrix 80/100 Chromosorb P NAW support, L × O.D. × I.D. 1.3 ft (0.40 m) × 1/8  in. × 2.1 mm

  •  NACRES SB.54

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Metal Packed GC Columns (General Configuration), Packed GC Columns, Packed GC Columns and Components More...
Quality Level   100
material   stainless steel column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   1.3 ft (0.40 m) × 1/8  in. × 2.1 mm
matrix   80/100 Chromosorb P NAW support
matrix active group   30% DC-200 (500 cstks) phase
column type   packed GC

Description

General description

Packed column made up of stainless steel are basically used when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents.

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column is of a general configuration, and can be carefully bent to fit most instruments.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?