• USA Home
  • 20073-U - ORBO 306 Petroleum Charcoal (20/40), 400/200 mg

EMAIL THIS PAGE TO A FRIEND
20073-U Supelco

ORBO 306 Petroleum Charcoal (20/40), 400/200 mg

W,F,F separators, O.D. × L 8 mm × 110 mm, pkg of 50 tubes

Synonym: Petroleum Charcoal Tube

  •  NACRES NB.24

Purchase

Properties

Related Categories Active Sampling, Air Monitoring, Analytical/Chromatography, Charcoal Sorbent Tubes, ORBO Sorbent Tubes For Gas & Vapor Sampling,
Quality Level   100
material   W,F,F separators
composition   Bed A, 400 mg
  Bed B, 200 mg
mfr. no.   ORBO 306
packaging   pkg of 50 tubes
application(s)   air monitoring: suitable
O.D. × L   8 mm × 110 mm
matrix   Petroleum charcoal
  petroleum charcoal charcoal
particle size   20-40 mesh
  20-40 mesh
Featured Industry   Environmental
Industrial Hygiene
compatibility   mode of use Solvent desorption
  technique used for Air monitoring

Description

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?