• USA Home
  • 20242 - Carbotrap® 200 Dynatherm 890, 850

EMAIL THIS PAGE TO A FRIEND
20242 Supelco

Carbotrap® 200 Dynatherm 890, 850

for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU and OI Analytical Air Tube Desorber or DMP-16

Purchase

Properties

Related Categories Active Sampling, Air Monitoring, Analytical/Chromatography, Dynatherm Standard Sampling Tubes, Sample Preparation & Purification,
material   glass TD tube (non-fritted)
feature   preconditioned
composition   Carbosieve S-III
  Glass beads
packaging   pkg of 1 ea
mfr. no.   Supelco Carbotrap® 200
I.D. × O.D.   4 mm × 6 mm
matrix   Carbotrap® 200
Featured Industry   Environmental
Industrial Hygiene
compatibility   for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU and OI Analytical Air Tube Desorber or DMP-16

Description

Application

C2-C14 compounds in air

Legal Information

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC

Dynatherm is a trademark of CDS Analytical

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Thermal Desorption Tube Selection Guide

In addition to the general guidelines presented in the tables below view the technical report titled “A Tool for Selecting an Adsorbent for Thermal Desorption Applications” (1,770Kb pdf - T402025), w...
Keywords: Sample preparations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?