• USA Home
  • 20924 - Empty Tube for Thermal Desorption
EMAIL THIS PAGE TO A FRIEND
20924 Supelco
Empty Tube for Thermal Desorption
stainless steel TD tube, O.D. × I.D. × L 1/2  in. × 12 mm × 7 in., pkg of 1 ea
Synonym: Tekmar® Empty SS TD Tube
Purchase
Properties
Related Categories Active Sampling, Air Monitoring, Analytical/Chromatography, Sample Preparation & Purification, Tekmar Thermal Desorption Tubes (Stainless Steel, 1/2″ OD X 7″ Long, 12 mm ID),
material   stainless steel TD tube
packaging   pkg of 1 ea
O.D. × I.D. × L   1/2  in. × 12 mm × 7 in.
Featured Industry   Environmental
Industrial Hygiene
compatibility   for use with Tekmar AEROTrap 6000 models
Description
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
Thermal Desorption Tube Selection Guide
In addition to the general guidelines presented in the tables below view the technical report titled “A Tool for Selecting an Adsorbent for Thermal Desorption Applications” (1,770Kb pdf - T402025), w...
Keywords: Sample preparations
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?