• USA Home
  • 21101AST - Astec® CYCLOBOND® I 2000 AC Chiral HPLC Guard

EMAIL THIS PAGE TO A FRIEND
21101AST Supelco

Astec® CYCLOBOND® I 2000 AC Chiral HPLC Guard

Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, Astec CYCLOBOND Chiral HPLC Columns (Native and Derivatized Cyclodextrins), CYCLOBOND I 2000 AC, Chiral HPLC Columns, Chiral HPLC Guard Columns and Accessories,
description   HPLC guard column
packaging   pkg of 1 ea
application(s)   HPLC: suitable
L × I.D.   2 cm × 4 mm
matrix   silica particle platform
matrix active group   cyclodextrin, beta- phase
particle size   5 μm
pore size   100 Å
separation technique   chiral

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC

CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?