• USA Home
  • 21543 - SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
EMAIL THIS PAGE TO A FRIEND
21543
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 10 μL, 10F-HP-0.63, needle size 23 ga (cone tip), needle L × O.D. 42 mm × 0.63 mm
Purchase
Properties
Related Categories Agilent 7673/7683, Analytical/Chromatography, GC Autosampler Syringes, SGE Color-Coded Syringes for Agilent / HP 7673 & 7683 Autosampler, SGE MicroVolume syringes for autosamplers,
material   cone tip stainless steel needle (point style 1)
needle size   23 ga (cone tip)
feature   needle type F (fixed)
packaging   pkg of 1 ea
needle L × O.D.   42 mm × 0.63 mm
volume   10 μL
compatibility   for use with Agilent/HP 7673 and 7683 autosampler
Description
General description
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?