• USA Home
  • 21765 - Empty Glass GC Column, for Varian® 3000, 4000, 6000, and Vista (FID, Inj. A to Det. A, On-Column)
EMAIL THIS PAGE TO A FRIEND
21765 Supelco
Empty Glass GC Column, for Varian® 3000, 4000, 6000, and Vista (FID, Inj. A to Det. A, On-Column)
L × O.D. × I.D. 8.6 ft (2.6 m) × 1/4  in. × 2.0 mm
Purchase
Properties
Related Categories Analytical/Chromatography, Empty GC Columns, Empty Glass GC Columns, Empty Glass GC Columns, for Varian 3000, 4000, 6000, and Vista (FID, inj A to det A), Gas Chromatography (GC),
material   glass column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   8.6 ft (2.6 m) × 1/4  in. × 2.0 mm
column type   empty
Description
General description
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.
Silane treated empty glass column. Fittings are not included.
• X (length of injector arm) = 9.31 in. (236 mm)
• Y (length of detector arm) = 8.0 in. (203 mm)
• S (span, injector to detector) = 5.5 in. (140 mm)
This column fits Varian 3000, 4000, 6000, and Vista (FID, injector A to detector A, on-column injection).
Application
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Legal Information
Varian is a registered trademark of Varian Medical Systems, Inc.
Vista is a trademark of Varian Medical Systems, Inc.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?