• USA Home
  • 22022-U - Swagelok® Front Ferrule

EMAIL THIS PAGE TO A FRIEND
22022-U

Swagelok® Front Ferrule

Swagelok®, 103-1, brass, 1/16 in. Swagelok

Synonym: Swagelok® 103-1,BRASS FRT FERR 1/16" PK10

  •  NACRES SB.54

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation, Swagelok Front Ferrule,
material   brass
packaging   pkg of 10 ea
mfr. no.   Swagelok®, 103-1
fitting   1/16 in. Swagelok

Description

General description

Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing.

Application

Brass front ferrule maybe used for sealing the tube ends to increase the lifetime and performance of the permeation tubes as standard gas sources.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?