• USA Home
  • 22024AST - Astec® CYCLOBOND® I 2000 Chiral HPLC Column

EMAIL THIS PAGE TO A FRIEND
22024AST Supelco

Astec® CYCLOBOND® I 2000 Chiral HPLC Column

10 μm particle size, L × I.D. 25 cm × 4.6 mm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, Astec CYCLOBOND Chiral HPLC Columns (Native and Derivatized Cyclodextrins), CYCLOBOND I 2000, Chiral HPLC Columns, HPLC/UHPLC More...
packaging   pkg of 1 ea
mfr. no.   Astec
parameter   0-50 °C temperature
  172 bar pressure (2500 psi)
application(s)   HPLC: suitable
L × I.D.   25 cm × 4.6 mm
matrix   silica particle platform
matrix active group   cyclodextrin, beta- phase
particle size   10 μm
pore size   100 Å
separation technique   chiral

Description

General description

CYCLOBOND I 2000 is bonded with β-cyclodextrin by a patented process to produce a stable matrix with the cyclodextrin arranged in such a way as to retain its most valuable property of forming inclusion complexes. This allows them to effect numerous chemical separations by selectively including into their cavities a wide variety of organic molecules. Non-inclusion type separations are also possible with the polar organic mode for a wide variety of molecule types.

• Bonded phase: Underivatized, native β-cyclodextrin

Resources:
CYCLOBOND FAQs
CYCLOBOND Reference Bibliography
Chiral Product Literature

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC

CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Analysis of Isoprenoid Pathway Metabolites by LC-MS | Biofiles, Vol. 8, No. 17

Isoprenoid pathway metabolites such isoprenoidphosphates and-pyrophosphates are central biological intermediates leading to sterols, dolichols, ubiquinones, prenylated natural products and proteins. ...
Rudolf Köhling, Roland Meier, Bernhard Schönenberger and Roland Wohlgemuth

Sigma-Aldrich®, Buchs, Switzerland
Biofiles, Vol. 8, No. 17
Keywords: Buffers, Chromatography, High performance liquid chromatography, Individual protein Immunoprecipitation, Mass spectrometry, Metabolites, Separation, Solvents

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?