• USA Home
  • 22072 - Swagelok® Reducing Union

EMAIL THIS PAGE TO A FRIEND
22072 Supelco

Swagelok® Reducing Union

Swagelok®, 300-6-2, brass, 3/16 in. Swagelok

Synonym: Swagelok® 300-6-2,BR.REDUC.UNION 3/16X1/8 PK2

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation, Swagelok Reducing Union,
material   brass
packaging   pkg of 2 ea
mfr. no.   Swagelok®, 300-6-2
fitting   1/8 in. Swagelok
  3/16 in. Swagelok

Description

General description

Nuts and ferrules included.

Reducing unions of Swagelok® type are mostly used as metal column terminators. All original reducing unions have a bore in the centre of about 6mm in length and an internal diameter of 1.2mm. Moreover, reducing unions have a large bore at the non-reducing site. Modification is necessary in reducing union in order to minimize peak broadening, pressure resistance and to ease the installation process.

Application

Brass Swagelok® reducing union may be used for providing connections for the glass tubing fitted in it to the stainless steel tubing by means of PTFE ferrules, in an elution technique used for the determination of solubility of gases like argon, helium, nitrogen and carbon dioxide in molten salts at pressures ranging between 0.5 to 2atm.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?