• USA Home
  • 22447 - Packed Column Filling Kit
EMAIL THIS PAGE TO A FRIEND
22447 Supelco
Packed Column Filling Kit
Purchase
Description
General description
This convenient kit contains many of the items need to quickly and efficiently pack columns. Includes two puller/inserter tools (Cat. No. 22406), plastic funnel, rubber column connector, and 50 g of silanized glass wool for plugs.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?