• USA Home
  • 22552 - Vinyl chloride in nitrogen

EMAIL THIS PAGE TO A FRIEND
22552 Supelco

Vinyl chloride in nitrogen

100 ppm, cylinder of 14 L (stainless steel), analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Organic Pollutant Standards, Q-Z, Volatile & Semivolatile Standards More...
grade   analytical standard
form   gas
packaging   cylinder of 14 L (stainless steel)
mfr. no.   Scott Gas, T73-14
availability   not available in EU, Canada
concentration   100 ppm
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   multi-component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

• Vinyl chloride in nitrogen, 100 ppm is a two component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.
• A certificate of analysis is included.
• Packaged in lightweight, non-refillable, recyclable containers.
• Portable, convenient, and easy to use.
• Manufactured gravimetrically for maximum accuracy.
• NIST traceable.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1956 2.2
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

regulator

Product #

Description

Add to Cart

507911 Model 24 Single-Stage Regulator, for use with CGA-160 for SCOTTY 14 cylinder (This item does not include the syringe adapter).

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?