EMAIL THIS PAGE TO A FRIEND
22660-U Supelco

Leak Tester Kit

  •  NACRES SB.54

Purchase

Description

General description

This kit allows septum leaks to be detected without the risk of contaminating the system. Simply dip one end of a leak tester tube into SNOOP®, then place the other end over the septum nut or needle guide. Bubbles indicate a leak. Kit includes 10 leak tester tubes and 8 ounces of SNOOP.

Legal Information

SNOOP is a registered trademark of Swagelok Company

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?