• USA Home
  • 23381 - Mixed Cellulose Ester (MCE) Membrane Filter
EMAIL THIS PAGE TO A FRIEND
23381 Supelco
Mixed Cellulose Ester (MCE) Membrane Filter
GN-4 Metricel® membrane, pore size 0.8 μm, O.D. 37 mm
Purchase
Properties
Related Categories Active Sampling, Air Monitoring, Air Monitoring Cassettes & Membrane Filters, Analytical/Chromatography, Membrane Filters,
Quality Level   100
material   GN-4 Metricel® membrane
Agency/Method   ASTM® D4185,D4856
  NIOSH 5004,5009,5011,5020,5025,5026,5033,5034,5037,5038,5039,5508,5510,5512,5514,5519,6007,7013,7020,7024,7027,7029,7030,7048,7056,7074,7082,7102,7105,7300,7601,7602,7700,7701,7702,7900,7904
  OSHA ID-104,ID-110,ID-113,ID-111,ID-125G,ID-121,ID-120,ID-145,ID-189,ID-206,ID-204,ID-213,ID-211
description   (MCE, with pad)
packaging   pkg of 100 ea
O.D.   37 mm
pore size   0.8 μm
Featured Industry   Environmental
Industrial Hygiene
Description
General description
• Mixed cellulose ester (MCE) membrane filters are accepted for air monitoring of fibers, asbestos fibers and airborne metals such as lead (NIOSH Methods 7400 and 7402).
• It can also be used to monitor respirable particulates such as silica, metal and dusts.
• 0.8 mm GN-4 Metricel (mixed cellulose esters) membrane filters have a low fiber background count.
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
Metricel is a registered trademark of Pall Corporation
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?