• USA Home
  • 23999 - QC+ Point-of-Operation Panel
EMAIL THIS PAGE TO A FRIEND
23999 Supelco
QC+ Point-of-Operation Panel
3-head QC+ panel, fitting size 1/8 in.
Synonym: QC+ POINT OF OPERATION PANEL, 3-HEAD, 1/8IN, W/CARTRIDGES
Purchase
Properties
feature   3-head QC+ panel
fitting size   1/8 in.
Description
General description
QC+ panels make cartridge replacement fast and easy. Rotate the valve handle(s) on the filter head(s) counterclockwise, to allow gas to pass through the head. Replace the cartridge, then rotate the valve handle clockwise, to puncture the seal on the new cartridge and restore gas flow through the purifier bed.
• Replace up to four cartridges in one minute or less; eliminate purge time
The 3-Head QC+ Panel comes complete with:
• GC-3 Hydrocarbon-Removing cartridge
• GC-2 Water-Removing cartridge
• GC-1 Oxygen-Removing cartridge
The 4-Head QC+ Panel comes complete with:
• GC-3 Hydrocarbon-Removing cartridge
• GC-2 Water-Removing cartridge
• GC-1 Oxygen-Removing cartridge
• GC-4 Indicating Oxygen-Removing cartridge
Replacement cartridges provide additional options:
The SupelCarb® Hydrocarbon-Removing cartridge has twice the capacity for hydrocarbons, relative to the activated charcoal in the GC-3 Hydrocarbon-Removing cartridge.
The GC2-1 Indicating Water-Removing cartridge contains a mixture of Molecular Sieve 5A and Drierite®. The color changes from blue to pink as the material becomes spent.
The GC-4 Indicating Oxygen-Removing cartridge contains manganese oxide. The color changes from green to brown as the material becomes spent.
Legal Information
Supelcarb is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?