• USA Home
  • 24080UINT - SUPELCOWAX® 10 Intuvo Capillary GC Column

EMAIL THIS PAGE TO A FRIEND
24080UINT Supelco

SUPELCOWAX® 10 Intuvo Capillary GC Column

column L × I.D. 30 m × 0.32 mm, df 0.25 μm, for use with Agilent Intuvo GC

Synonym: SUPELCOWAX® 10 Intuvo Capillary GC Column

Purchase

Properties

df   0.25 μm
column L × I.D.   30 m × 0.32 mm
compatibility   for use with Agilent Intuvo GC

Description

Legal Information

SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Gas Chromatography Columns for the Agilent® Intuvo GC System

Available now! Get your favorite Supelco® GC columns for use with the Agilent® Intuvo GC System!
Keywords: Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?