• USA Home
  • 24104UINT - SP®-2331 Intuvo Capillary Column

EMAIL THIS PAGE TO A FRIEND
24104UINT Supelco

SP®-2331 Intuvo Capillary Column

L × I.D. 60 m × 0.25 mm, for use with Agilent Intuvo GC

Synonym: SP®-2331 Capillary GC Column

Purchase

Properties

material   fused silica
df   0.2 μm
L × I.D.   60 m × 0.25 mm
matrix active group   Stabilized; proprietary phase
column type   capillary highly polar
compatibility   for use with Agilent Intuvo GC

Description

General description

Application: A highly polar cyanosiloxane column specially tested for analyses of dioxins, specifically tetrachlorodibenzodioxin (TCDD) isomers. Because the phase is stabilized, it has a maximum temperature slightly higher than non-bonded cyanosiloxane columns.
USP Code: None
Phase:
• Stabilized
• Proprietary
Temp. Limits:
• Subambient to 275 °C (isothermal or programmed)

Legal Information

SP is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Gas Chromatography Columns for the Agilent® Intuvo GC System

Available now! Get your favorite Supelco® GC columns for use with the Agilent® Intuvo GC System!
Keywords: Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?