• USA Home
  • 24424 - SGE Needles for RN syringes
EMAIL THIS PAGE TO A FRIEND
24424
SGE Needles for RN syringes
volume 0.5 μL, needle size 23 ga (cone tip), needle L × O.D. 50 mm × 0.63 mm
Synonym: REPL.NEEDLE NP0.5BN-5C, EA
Purchase
Properties
Related Categories Analytical/Chromatography, GC Manual Syringes, Liquid sampling, SGE Needles, SGE Replacement Kits for plunger-in-needle syringes,
material   cone tip stainless steel needle (point style 1)
needle size   23 ga (cone tip)
description   NP 0.5BN-5C
feature   cone tip (point style 1)
packaging   pkg of 1 ea
mfr. no.   SGE, 033010
needle L × O.D.   50 mm × 0.63 mm
volume   0.5 μL
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?