• USA Home
  • 25727 - Intermediate Polar Fused Silica Tubing

25727 Supelco

Intermediate Polar Fused Silica Tubing

L × I.D. 3.0 m × 0.25 mm



Related Categories Analytical/Chromatography, Capillary GC Columns and Guard Columns/Retention Gaps, Fused Silica Tubing, Gas Chromatography (GC), Guard Columns/Retention Gaps,
composition   phenyl/methyl (deactivated)
packaging   pkg of 1 ea
parameter   360 °C max. temp.
application(s)   gas chromatography (GC): suitable
L × I.D.   3.0 m × 0.25 mm

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

How to Choose a Capillary GC Column

An optimized chromatographic separation begins with the column. The selection of the proper capillary column for any application should be based on four significant factors: stationary phase, column ...
How to Choose a Capillary GC Column
Keywords: Chromatography, Condensations, Gas chromatography, Immobilization, Mass spectrometry, Pesticides, Separation, Size-exclusion chromatography, Solid phase microextractions, Solvents, Vaporization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?