• USA Home
  • 25965 - Glass Packed GC Column (fits Agilent, Configuration "A")

25965 Supelco

Glass Packed GC Column (fits Agilent, Configuration "A")

phase 1.5% SP-2250 + 1.95% SP-2401, matrix 100/120 SUPELCOPORT support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 4.0 mm



Related Categories Analytical/Chromatography, Gas Chromatography (GC), Glass Packed GC Columns (fit Agilent, Configuration "A"), Packed GC Columns, Packed GC Columns and Components More...
material   glass column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   6.0 ft (1.8 m) × 1/4 in. × 4.0 mm
matrix   100/120 SUPELCOPORT support
matrix active group   1.5% SP-2250 + 1.95% SP-2401 phase
Agency/Method   EPA 608 (pesticides)
  NIOSH 5503 (Aroclors)
column type   packed GC


General description

Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?