• USA Home
  • 26051 - Glass Packed GC Column (fits PerkinElmer®)

EMAIL THIS PAGE TO A FRIEND
26051 Supelco

Glass Packed GC Column (fits PerkinElmer®)

phase 5% Carbowax 20M, matrix 60/80 Carbopack B support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4  in. × 2.0 mm

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Glass Packed GC Columns (fit PerkinElmer), Packed GC Columns, Packed GC Columns and Components More...
material   glass column
mfr. no.   PerkinElmer
parameter   0-225 °C temperature (isothermal or programmed)
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   6.0 ft (1.8 m) × 1/4  in. × 2.0 mm
matrix   60/80 Carbopack B support
matrix active group   5% Carbowax 20M phase
column type   packed GC

Description

General description

Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits PerkinElmer 8000, 9000, AutoSystem, XL, and Clarus 500 (Not On-Column Injection).

Application

blood alcohols, solvents

Legal Information

PerkinElmer is a registered trademark of PerkinElmer Inc.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?