• USA Home
  • 2632605 - Inlet Liner, Split Type, Frit Design (unpacked)
EMAIL THIS PAGE TO A FRIEND
2632605 Supelco
Inlet Liner, Split Type, Frit Design (unpacked)
pkg of 5 ea
Purchase
Description
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
• Fits ATAS (Optic 2)
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
Changing the GC Inlet Liner
No sample is perfectly clean and over time nonvolatile sample residues contaminate liners. As these contaminants accumulate they adversely affect chromatography by creating sites that lead to the abs...
Keywords: Absorption, Chromatography, Gas chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?