EMAIL THIS PAGE TO A FRIEND
27074-U Supelco

Vials, crimp top

volume 0.1 mL, plastic vial (with glass inner cone), vial type, conical bottom, Snap Ring/Crimp top, pkg of 100 ea

  •  NACRES NB.11

Purchase

Properties

Related Categories 11 mm crimp seal, Analytical/Chromatography, Crimp Top Vials, 12 x 32 mm, Crimp Top Vials, Standard Opening, 12 x 32 mm, Crimp top,
material   plastic vial (with glass inner cone)
feature   closure type crimp top vial
  vial type conical bottom
packaging   pkg of 100 ea
O.D. × H × I.D.   12 mm × 32 mm × 4.6 mm
volume   0.1 mL
fitting   thread for 11 mm
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

27214-U Crimp seals with PTFE/silicone septa, silver aluminum seal, size × thickness 11 mm × 1.0 mm, white PTFE/silicone septum, pkg of 100 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?