• USA Home
  • 27080-U - Vials, screw top, clear glass

EMAIL THIS PAGE TO A FRIEND
27080-U Supelco

Vials, screw top, clear glass

marking spot, volume 2 mL, vial O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, thread for 8-425, pkg of × 100 ea

  •  NACRES NB.11

Purchase

Properties

Related Categories 2 mL Screw Top Vials, Standard Opening 4.6 mm, 201/202, 205 Series, 300 Series, 221/222, 231/401, 232/402,
More...
material   clear glass vial (with marking spot)
description   cap not included
feature   closure type screw top vial
  marking spot
packaging   pkg of × 100 ea
vial O.D. × H × I.D.   12 mm × 32 mm × 4.6 mm
volume   2 mL
fitting   thread for 8-425

Description

General description

Vials are suitable basically to inject samples from an autosampler. They are available in various sizes, although 2 mL vials are more commonly used. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. Screw top vials (marking spot, volume 2 mL, vial O.D. × H. 12 mm × 32 mm) are used on many autosamplers with a standard opening of 4.6 mm. The threads on the standard vials are 8-425.

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

23207 Space Saver storage racks, for use with 11-12 mm diameter vial (50 vials per rack), well opening 12.5 mm, pkg of 5 ea
27261-U Assembled screw cap with hole with PTFE/silicone septum, black polypropylene hole cap, thread for 8-425, PTFE/silicone, Top Hat (septa and closure designed to fit together tightly), for use with 2 mL vial, 4.6 mm opening, pkg of 100 ea
27262 Assembled screw cap with hole with PTFE/silicone septum, black polypropylene hole cap, 8-425 thread, PTFE/silicone, septum thickness 1.5 mm, for use with 2 mL vial (standard opening), pkg of 100 ea
27400-U Insert for 2 mL standard opening vial, 4.6 mm I.D., volume 0.05 mL, conical clear glass, bottom spring, O.D. × H 4 mm × 25 mm, pkg of 100 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?