• USA Home
  • 27089-U - Vials, screw top with phenolic open-top cap, pre-assembled
EMAIL THIS PAGE TO A FRIEND
27089-U Supelco
Vials, screw top with phenolic open-top cap, pre-assembled
volume 40 mL, clear glass vial, O.D. × H 27.5 mm × 95 mm, tan PTFE/silicone septum
  •  NACRES NB.11
Purchase
Properties
Related Categories Analytical/Chromatography, Autosampler Vials, Storage Kits and Accessories, Clear glass, Phenolic cap, PTFE/silicone septa, Pre-Assembled Sampling Vials,
material   black phenolic hole cap (open-top)
  clear glass vial
  tan PTFE/silicone septum
feature   closure type black phenolic hole cap
  closure type screw top vial
packaging   pkg of 100 ea
O.D. × H   27.5 mm × 95 mm
volume   40 mL
fitting   thread for 24-400
Description
General description
Vials (volume 40 mL, clear glass, O.D. × H. 27.5 mm × 95 mm) are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. The threads on the standard vials are 24-400. The phenolic cap is autoclavable and it can withstand the temperature up to 149° C. The tan PTFE/silicone septum is sized to fit screw top vials. It is suitable for use with most organics (until pierced) and is not recommended for use with chlorosilanes.
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
23206 Space Saver storage racks, for use with 29 mm diameter vial (50 vials per rack), well opening 30.0 mm, pkg of 5 ea
replacement part
Product #
Description
Add to Cart
27187 Screw cap, phenolic, with open center, black phenolic hole cap, for use with 20 or 40 mL vial with 24-400 thread, pkg of 100 ea
27188-U Septa, tan PTFE/silicone, white tan PTFE/silicone, diam. × thickness 22 mm × 0.100 in., for use with 20, 40 or 60 mL vial, pkg of 100 ea
27379 Vials, screw top, clear glass (vial only), volume 40 mL, clear glass vial, thread for 24-400, pkg of 100 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?