• USA Home
  • 27233 - Septa, PTFE/red rubber, pkg 100

EMAIL THIS PAGE TO A FRIEND
27233 Supelco

Septa, PTFE/red rubber, pkg 100

PTFE/red rubber, diam. × thickness 20 mm × 0.125 in., for use with 20 mm crimp seal, 22 mm screw cap

Purchase

Properties

Related Categories 20 mm, 22 mm diameter, fits 20, 40, 60 mL Screw Thread Vials, Analytical/Chromatography, Autosampler Vials, Storage Kits and Accessories, Material,
material   PTFE septum face
  PTFE/red rubber
  PTFE/red rubber
feature   temp. limit 120 °C
packaging   pkg of 100 ea
diam. × thickness   20 mm × 0.125 in.
compatibility   for use with 20 mm crimp seal, 22 mm screw cap

Description

General description

The red rubber is "polybutyl isoprene."

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

27016 Aluminum crimp seal (without septum), silver aluminum, tear-away, diam. 20 mm, pkg of 100 ea
27200 Aluminum crimp seal (without septum), silver aluminum, pressure release, diam. × opening 20 mm × 8 mm, pkg of × 100 ea
27227-U Aluminum crimp seal (without septum), silver aluminum, removable center, diam. 20 mm, pkg of 100 ea
27230-U Aluminum crimp seal (without septum), silver aluminum, open center, diam. × opening 20 mm × 9.5 mm, pkg of 100 ea
33183 Hand Decapper, for use with 20mm crimp seals, pkg of 1 ea
33280-U Crimpers, Hand crimper, for use with 20mm seals, pkg of 1 ea
22316-U Hand crimper, adjustable, Hand crimper, adjustable, for use with 20 mm crimp seals, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?