• USA Home
  • 27240-U - Crimp top vial convenience kit with PTFE/red rubber septa

EMAIL THIS PAGE TO A FRIEND
27240-U Supelco

Crimp top vial convenience kit with PTFE/red rubber septa

volume 2 mL, clear glass vial, vial O.D. × H × I.D. 12 mm × 32 mm × 6 mm, PTFE/red rubber aluminum seal, pkg of 1000 ea

  •  NACRES NB.11

Purchase

Properties

Related Categories Analytical/Chromatography, Crimp Top Vials, 12 x 32 mm, Crimp top vials, Crimp top, 12 x 32 mm, convenience kits, large opening, Crimp vials with PTFE/rubber seals,
material   PTFE/red rubber aluminum seal
  clear glass vial
  silver aluminum seal
feature   closure type crimp top vial
packaging   pkg of 1000 ea
seal   11 mm
vial O.D. × H × I.D.   12 mm × 32 mm × 6 mm
volume   2 mL
fitting   thread for 11 mm
Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?