• USA Home
  • 28077-U - Equity®-1 Capillary GC Column

EMAIL THIS PAGE TO A FRIEND
28077-U Supelco

Equity®-1 Capillary GC Column

L × I.D. 60 m × 0.53 mm, df 3.00 μm

  •  eCl@ss 32119290

  •  NACRES SB.54

Purchase

Properties

Related Categories All Gas Chromatography Columns, Analytical/Chromatography, Capillary GC Columns, Capillary GC Columns, by name, Equity-1,
Quality Level   100
material   fused silica
Agency/Method   ASTM® D1945,D3710,D4815,D5442,D5580,D7096
  EN 14110
  EPA 8015,TO-17
  GPA 2186,2286
  NIOSH 1403,1500/1501,2542
  OSHA 53,56
  meets requirements for USP G1, G2, and G9
parameter   -60-260 °C temperature (isothermal)
  -60-280 °C temperature (programmed)
Beta value   44
df   3.00 μm
application(s)   gas chromatography (GC): suitable
L × I.D.   60 m × 0.53 mm
matrix active group   Bonded; poly(dimethyl siloxane) phase
Featured Industry   Agriculture
Biofuels
Chemicals and Industrial Polymers
Cleaning Products
Clinical
Cosmetics
Flavors and Fragrances
Food and Beverages
Forensics and Toxicology
Industrial Hygiene
Personal Care
Petroleum
column type   capillary non-polar

Description

General description

Application: This column is designed for general purpose applications where a non-polar column is required. Analytes will be separated primarily according to boiling point.
USP Code: This column meets USP G1, G2, and G9 requirements.
Phase:
• Bonded
• Poly(dimethyl siloxane)
Temp. Limits:
• ≤0.32 mm I.D., <2 μm: -60 °C to 325 °C (isothermal) or 350 °C (programmed)
• ≤0.32 mm I.D., ≥2 μm: -60 °C to 300 °C (isothermal or programmed)
• ≥0.53 mm I.D., <2 μm: -60 °C to 300 °C (isothermal) or 320 °C (programmed)
• ≥0.53 mm I.D., ≥2 μm: -60 °C to 260 °C (isothermal) or 280 °C (programmed)

Other Notes

Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Equity is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

How to Choose a Capillary GC Column

An optimized chromatographic separation begins with the column. The selection of the proper capillary column for any application should be based on four significant factors:
How to Choose a Capillary GC Column
Keywords: Chromatography, Condensations, Gas chromatography, Immobilization, Mass spectrometry, Pesticides, Separation, Size-exclusion chromatography, Solid phase microextractions, Solvents, Vaporization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?