• USA Home
  • 2876701-U - Inlet Liner, Splitless Type, Straight Design (unpacked)

EMAIL THIS PAGE TO A FRIEND
2876701-U Supelco

Inlet Liner, Splitless Type, Straight Design (unpacked)

Split for ThermoQuest 8000 and TRACE, staraight through, pkg of 1 ea

  •  NACRES SB.54

Purchase

Description

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
• Fits Thermo (ThermoQuest 8000 and TRACE)

Legal Information

TRACE is a trademark of Thermo Fisher Scientific or its subsidiaries

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Changing the GC Inlet Liner

No sample is perfectly clean and over time nonvolatile sample residues contaminate liners. As these contaminants accumulate they adversely affect chromatography by creating sites that lead to the abs...
Keywords: Absorption, Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?