• USA Home
  • 2877405-U - Inlet Liner, Splitless Type, Single Taper Design (unpacked)

EMAIL THIS PAGE TO A FRIEND
2877405-U Supelco

Inlet Liner, Splitless Type, Single Taper Design (unpacked)

pkg of 5 ea

  •  NACRES SB.54

Purchase

Properties

Related Categories Analytical/Chromatography, GC Accessories, Gas Chromatography (GC), Inlet Liners for Thermo (ThermoQuest 8000 and TRACE), Inlet Liners, Glass Wool, and Specialized Hand Tools,
description   Splitless for ThermoQuest 8000 and TRACE with taper
feature   Splitless with Single Taper
packaging   pkg of 5 ea
L × O.D. × I.D.   105 mm × 8.0 mm × 3.0 mm
  105 mm × 8.0 mm × 3.0 mm
compatibility   configured for Finnigan ThermoQuest 8000 and Trace

Description

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
• Fits Thermo (ThermoQuest 8000 and TRACE)

Legal Information

TRACE is a trademark of Thermo Fisher Scientific or its subsidiaries

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Changing the GC Inlet Liner

No sample is perfectly clean and over time nonvolatile sample residues contaminate liners. As these contaminants accumulate they adversely affect chromatography by creating sites that lead to the abs...
Keywords: Absorption, Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?