• USA Home
  • 2877701-U - Inlet Liner, Splitless Type, Middle Gooseneck Design (unpacked)

EMAIL THIS PAGE TO A FRIEND
2877701-U Supelco

Inlet Liner, Splitless Type, Middle Gooseneck Design (unpacked)

pkg of 1 ea

  •  NACRES SB.54

Purchase

Description

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
• Fits Shimadzu (14, 15A, and 16) [with SPL-14 Injector]

Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Changing the GC Inlet Liner

No sample is perfectly clean and over time nonvolatile sample residues contaminate liners. As these contaminants accumulate they adversely affect chromatography by creating sites that lead to the abs...
Keywords: Absorption, Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?