• USA Home
  • 2878101-U - Inlet Liner, Split/Splitless Type, Straight FocusLiner Design (wool packed)
EMAIL THIS PAGE TO A FRIEND
2878101-U Supelco
Inlet Liner, Split/Splitless Type, Straight FocusLiner Design (wool packed)
pkg of 1 ea
Purchase
Description
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
• Fits Shimadzu (14, 15A, and 16) [with SPL-14 Injector]
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Articles
Changing the GC Inlet Liner
No sample is perfectly clean and over time nonvolatile sample residues contaminate liners. As these contaminants accumulate they adversely affect chromatography by creating sites that lead to the abs...
Keywords: Absorption, Chromatography, Gas chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?