• USA Home
  • 29061-U - Vials, 2 mL, open-top screw cap, R.A.M. (9 mm) convenience kit with closures with septa, unassembled, pkg of 100

EMAIL THIS PAGE TO A FRIEND
29061-U Supelco

Vials, 2 mL, open-top screw cap, R.A.M. (9 mm) convenience kit with closures with septa, unassembled, pkg of 100

volume 2 mL, amber glass vial, PTFE/red rubber septum

  •  NACRES NB.11

Purchase

Description

Suitability

Autosampler compatibility:
Agilent
Agilent 1050/1090A
Agilent 1100/1200
Agilent 7673A, 7683
Agilent 5890/7985A/6890
Hitachi
AS-1000, AS-2000, AS-4000
L7200/L7250
LEAP Technologies
CTC LC PAL, GC PAL
CombiPAL
HTC PAL/HTS PAL
PerkinElmer
Autosystem/AS-2000/XL
Clarus 500/600
ISS-100, 200
Shimadzu LC Autosamplers
SIL-HTa/SIL-HTc
SIL-10ADVP
SIL-10A/SIL-10Ai/SIL-10Ap
Varian
CP-8410
8100/8200
9100/90950/9095
Waters
Alliance 2690
Acquity
Breeze
717Plus

Other Notes

The diameter of inserts varies between manufacturers. Only the following insert part numbers are compatible with these large opening vials.
• 24719, 24720 Glass insert, conical bottom, 6 x 29mm
• 24721 Glass insert with bottom spring, 6 x 29mm
• 24722 Polypropylene insert with bottom spring, 6 x 29mm
• 24717, 24718 Glass insert, conical bottom, 6 x 31mm
• 24715, 24716 Glass insert, flat bottom, 6 x 31mm

Legal Information

R.A.M. is a trademark of J.G. Finneran Associates, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

23207 Space Saver storage racks, for use with 11-12 mm diameter vial (50 vials per rack), well opening 12.5 mm, pkg of 5 ea
24722 Inserts for 2 mL large opening vials, 6.0 mm I.D., volume 0.2 mL, conical polypropylene (with bottom spring), O.D. × H × I.D. 6 mm × 29 mm × 4.6 mm, pkg of 100 ea
24721 Inserts for 2 mL large opening vials, 6.0 mm I.D., volume 0.2 mL, conical clear glass (with bottom spring), O.D. × H × I.D. 6 mm × 29 mm × 4.6 mm, pkg of 100 ea
29004-U Vials, screw top, R.A.M. (9 mm thread), large opening, 12 x 32 mm, volume 2 mL, amber glass vial, pkg of × 100 ea
29043-U Screw cap (open-top), 9 mm thread, with liner, black polypropylene cap, red rubber/PTFE septum, pkg of 100 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?