• USA Home
  • 29121-U - Vials, open-top screw cap, 2 mL large opening with closures, unassembled convenience packs

EMAIL THIS PAGE TO A FRIEND
29121-U Supelco

Vials, open-top screw cap, 2 mL large opening with closures, unassembled convenience packs

volume 2 mL, amber glass vial, thread for 10-425, black polypropylene cap, PTFE/silicone/PTFE septa

  •  NACRES NB.11

Purchase

Properties

Related Categories 2 mL Screw Top Vials (12 x 32 mm), Large Opening (6 mm) Taking 10 mm Septa, 2 mL Screw Top Vials, Large Opening, Analytical/Chromatography, Convenience kits for screw thread vials, Convenience kits, 2 mL Large Opening Vials,
material   PTFE/silicone/PTFE septa
  amber glass vial
  black polypropylene cap
description   Unassembled
feature   closure type black polypropylene hole cap
  closure type screw top vial
packaging   pkg of × 100 ea
O.D. × H × I.D.   12 mm × 32 mm × 6 mm
volume   2 mL
fitting   thread for 10-425

Description

Suitability

Autosampler compatibility:
PerkinElmer Autosystem
PE ISS-100, 200, Integral 4000 and Series 200
Shimadzu GC and LC Autosamplers
Spark Marathon, Midas, Triathlon
Thermo HPLC Spectra System
Waters Alliance

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

23207 Space Saver storage racks, for use with 11-12 mm diameter vial (50 vials per rack), well opening 12.5 mm, pkg of 5 ea
24722 Inserts for 2 mL large opening vials, 6.0 mm I.D., volume 0.2 mL, conical polypropylene (with bottom spring), O.D. × H × I.D. 6 mm × 29 mm × 4.6 mm, pkg of 100 ea
27267-U Vials, screw top, amber glass (vial only), volume 2 mL, amber glass vial, thread for 10-425, pkg of 100 ea
24721 Inserts for 2 mL large opening vials, 6.0 mm I.D., volume 0.2 mL, conical clear glass (with bottom spring), O.D. × H × I.D. 6 mm × 29 mm × 4.6 mm, pkg of 100 ea
27407 Inserts for 2 mL large opening vials, 6.0 mm I.D., volume 0.25 mL, glass (with glass flange), for use with Step Vials, pkg of 100 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?