• USA Home
  • 29302-U - Certified Snap Ring CD vial kit with cap/septa, unassembled

EMAIL THIS PAGE TO A FRIEND
29302-U Supelco

Certified Snap Ring CD vial kit with cap/septa, unassembled

volume 1.5 mL, clear glass vial, snap top, PTFE/silicone, pkg of 100 ea

  •  NACRES NB.11

Purchase

Properties

Related Categories Analytical/Chromatography, Certified Autosampler Vials, Certified CD (Center Drain) Vials and Kits, 1.5 mL, Certified Center Drain (CD) Vials, Certified Center Drain (CD) vials,
material   PTFE/silicone
  blue polypropylene cap
  clear glass vial
feature   closure type blue polypropylene snap cap
  closure type snap ring vial
  cap is bonded to septa to prevent fallout
packaging   pkg of 100 ea
O.D. × H × I.D.   12 mm × 32 mm × 6 mm
volume   1.5 mL
fitting   for snap ring, 11 mm

Description

Legal Information

CD is a trademark of QIS, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

29305-U Certified polypropylene caps with septa, pkg of 100, snap cap, PTFE/silicone septum

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?