• USA Home
  • 29322-U - Certified CD vial kit, with Pico Pure Plus+ cap, unassembled

EMAIL THIS PAGE TO A FRIEND
29322-U Supelco

Certified CD vial kit, with Pico Pure Plus+ cap, unassembled

volume 1.5 mL, clear glass, 9 mm thread, white polyethylene cap, for use with LC/MS applications, pkg of 100 ea

  •  NACRES NB.11

Purchase

Properties

Related Categories Analytical/Chromatography, Certified CD (Center Drain) Vials and Kits, 1.5 mL, Certified Screw Thread Clear Glass Vial Kits, Certified Vials and Kits, Screw thread CD Vials,
material   clear glass
  white polyethylene cap
feature   closure type screw top vial
  closure type white polyethylene cap
packaging   pkg of 100 ea
O.D. × H × I.D.   12 mm × 32 mL × 6 mm
dead volume   <4 μL
volume   1.5 mL
fitting   thread for 9 mm screw thread, 9 mm
compatibility   for use with LC/MS applications

Description

General description

The CD Vial (Center Draining) has a unique conical interior bottom that allows the contents to completely drain to the center bottom of the vial. Maximum extractiion of the contents can be done utilizing either a micropipette or syringe.
The sturdy CD Vial with the patented unique is ideal for many micro applications.
The vials are manufactured from Boroisilicate Type 1 glass to safeguard against a change in the pH of the contents.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

CD is a trademark of QIS, Inc.

Pico Pure Plus+ is a trademark of AR Corp.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?