• USA Home
  • 29407-U - Certified crimp top vials, 12 x 32 mm

EMAIL THIS PAGE TO A FRIEND
29407-U Supelco

Certified crimp top vials, 12 x 32 mm

volume 2 mL, amber glass vial, pkg of 100 ea

  •  NACRES NB.11

Purchase

Description

Suitability

Autosampler compatibility:
Agilent
Agilent 1050/1090A
Agilent 1100/1200
Agilent 7673A, 7683
Agilent 5890/7985A/6890
Hitachi
AS-1000, AS-2000, AS-4000
L7200/L7250
LEAP Technologies
CTC LC PAL, GC PAL
CombiPAL
HTC PAL/HTS PAL
PerkinElmer
Autosystem/AS-2000/XL
Clarus 500/600
ISS-100, 200
Varian
CP-8410
8100/8200
9100/90950/9095

Other Notes

The inside diameter of vials vary with the manufacture. The following insert part numbers are compatible with these large opening vials.
• 29435-U PK100, Certified 200 μL glass inserts, mandrel precision point, 6 X 29mm, with bottom spring
• 29436-U PK100, Certified 250 μL glass insert, conical bottom, 6 x 31 mm
• 29437-U PK100, Certified 300 μL glass inserts, conical bottom, 6 x 30 mm
• 29441-U PK100, Certified 350 μL glass inserts, flat bottom, 6 x 31mm

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

29435-U Certified glass inserts for 12 x 32 mm, large opening vials, volume 0.2 mL, clear glass insert (with plastic bottom spring), O.D. × H 6 mm × 29 mm, pkg of 100 ea
29436-U Certified glass inserts for 12 x 32 mm, large opening vials, volume 0.25 mL, clear glass insert (no spring required), conical point, O.D. × H 6 mm × 31 mm, pkg of 100 ea
29437-U Certified glass inserts for 12 x 32 mm, large opening vials, volume 0.3 mL, clear glass insert, conical point, O.D. × H 6 mm × 31 mm, pkg of 100 ea
29441-U Certified glass inserts for 12 x 32 mm, large opening vials, volume 0.35 mL, clear glass insert (flat bottom), O.D. × H 6 mm × 31 mm, pkg of 100 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?