• USA Home
  • 29592-U - DIN1 Gas Cylinder Pressure Regulator

EMAIL THIS PAGE TO A FRIEND
29592-U

DIN1 Gas Cylinder Pressure Regulator

1-stage ultra high purity, parameter 0-100 psi

  •  NACRES SB.54

Purchase

Description

General description

For use with hydrogen, methane, or 5% methane in argon gas cylinders.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?