EMAIL THIS PAGE TO A FRIEND
29843-U

eVol Stand

Support stand, pkg of 1 ea

Synonym: eVOL® Components

  •  NACRES NB.03

Purchase

Properties

Related Categories Analytical/Chromatography, SGE eVol Analytical Electronic Syringe, Syringes and Syringe Accessories, eVol Stand
packaging   pkg of 1 ea
mfr. no.   SGE, 2910010

Description

Legal Information

eVol is a registered trademark of SGE Analytical Science Pty Ltd.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

29842-U eVol Electronic syringe, Electronic Syringe with programmable digital drive, pkg of 1 ea
29845-U eVol Single Changing Stand with adapters, Single charging stand with adapters, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?