• USA Home
  • 30042R - EPA Method 531.1 Analytes Mix

EMAIL THIS PAGE TO A FRIEND
30042R Supelco

EPA Method 531.1 Analytes Mix

100 μg/mL each component in methanol, analytical standard

Purchase

Properties

grade   analytical standard
packaging   ampule of 1 mL
concentration   100 μg/mL each component in methanol
Agency/Method   EPA 531.1
Featured Industry   Environmental
storage temp.   2-8°C

Analyte

Description

Product #

Add to Cart

Aldicarb    
Aldicarb-sulfone    
Aldicarb-sulfoxide    
Carbofuran    
Carbofuran-3-hydroxy    
1-Naphthyl-N-methylcarbamate    
Methiocarb    
Methomyl    
Oxamyl    
Propoxur    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1992 3/PG 2
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?